Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tris(1-chloro-2-propyl) phosphate | 13674-84-5 | MFCD00040408 | 98.0% | 327.57 g/mol | C9H18Cl3O4P | 100 G
SDP

Catalog No. 5000301585
Encompass_Preferred

Tris(1-chloro-2-propyl) phosphate (TCPP) is an organophosphate flame retardant supplied as a laboratory reagent for research applications. It is used in studies of flame retardancy, toxicology, and environmental fate, and is provided with supporting documentation for safe handling and characterization.

  • High purity (98.0%).
  • Molecular weight 327.57 g/mol.
  • Chemical formula C9H18Cl3O4P.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
  • Available in 100 g pack size.
  • Intended for research use only.

Catalog No. 50-003-01585 Supplier Medchemexpress LLC Supplier No. HYW023144100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris(1-chloro-2-propyl) phosphate (TCPP) is an organophosphate flame retardant supplied as a laboratory reagent for research applications. It is used in studies of flame retardancy, toxicology, and environmental fate, and is provided with supporting documentation for safe handling and characterization.

  • High purity (98.0%).
  • Molecular weight 327.57 g/mol.
  • Chemical formula C9H18Cl3O4P.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
  • Available in 100 g pack size.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.