Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(1-Pyrrolidinyl)Phosphine

Catalog No. 77544425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-544-425G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-544-425G Supplier Sigma Aldrich Supplier No. 7754442.5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Monodentate P-donor ligand.Used as a phosphitylation reagent for oligonucleotide synthesis.

Specifications

CAS 137846-38-9
Melting Point 240°C to 245°C
Molecular Formula C36H47Cl2N3O3Ru
Refractive Index n20/D 1.528; n20/D 1.53
Linear Formula C36H47Cl2N3O3Ru
Quantity 100 mg
Synonym Tripyrrolidinophosphine; Tris(N-pyrrolidinyl)phosphine; Tris(pyrrolidino)phosphine; Phosphorous acid tripyrrolidide; Tris(N,N-tetramethylene)phosphorous acid triamide
Molecular Weight (g/mol) 741.75 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.