Learn More
Bio Rad Laboratories TRIS 1KG
Catalog No. NC9683725
Encompass
Tris base (Tris[hydroxymethyl]aminomethane) is widely used in buffer preparation, including buffers for nucleic acid agarose, polyacrylamide gel electrophoresis, and blotting. Supplied as 1 kg. (161-0719)
Catalog No. NC9683725 Supplier Bio Rad Laboratories Supplier No. 1610719
Tris base (Tris[hydroxymethyl]aminomethane) is widely used in buffer preparation, including buffers for nucleic acid agarose, polyacrylamide gel electrophoresis, and blotting. Supplied as 1 kg. (161-0719)