Learn More
Strem, An Ascensus Company CAS# 14324-99-3. 1g. Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)manganese(III), 99% [Mn(TMHD)3]. MFCD00269825
SDP
Supplier:  Strem, An Ascensus Company 2550001G
Encompass
CAS# 14324-99-3. 1g. Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)manganese(III), 99% [Mn(TMHD)3]. MFCD00269825. Molecular Weight: 604.74. Molecular Formula: Mn(C11H19O2)3. Color/form: black xtl. Strem# 25-5000. http://www.strem.com/catalog/v/25-5000/
Catalog No. 50-138-5449