Learn More
Strem, An Ascensus Company CAS# 50525-27-4. 1g. Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate, min. 98%. MFCD00149670
SDP
Catalog No. 5090112037
Encompass
CAS# 50525-27-4. 1g. Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate, min. 98%. MFCD00149670. Molecular Weight: 640.54 (748.63). Molecular Formula: Ru(C10H8N2)3Cl2∙6H2O. Color/form: orange to red xtl. Strem# 44-7900. http://www.strem.com/catalog/v/44-7900/
Catalog No. 50-901-12037 Supplier Strem, An Ascensus Company Supplier No. 4479001G
CAS# 50525-27-4. 1g. Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate, min. 98%. MFCD00149670. Molecular Weight: 640.54 (748.63). Molecular Formula: Ru(C10H8N2)3Cl2∙6H2O. Color/form: orange to red xtl. Strem# 44-7900. http://www.strem.com/catalog/v/44-7900/