Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris[2-(2-methoxyethoxy)ethyl]amine

Catalog No. 301248500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-124-8500G 500 g
30-124-8100G 100 g
30-124-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 30-124-8500G Supplier Sigma Aldrich Supplier No. 301248500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tris[2-(2-methoxyethoxy)ethyl]amine was used:
  • In oxidation of arylmethanols under phase-transfer conditions
  • As cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7h-pyrrolo[2,3-d]pyrimidine
  • As phase-transfer catalyst in synthesis of 1,3-dideaza-2α-deoxyadenosine and related benzimidazole 2α-deoxyribonucleosides

Specifications

CAS 70384-51-9
Density 1.011 g/mL (at 25°C (literature))
Molecular Formula C15H33NO6
Linear Formula (CH3OCH2CH2OCH2CH2)3 N
Refractive Index n20/D 1.4486 (literature)
MDL Number MFCD00010748
Quantity 500 g
Synonym TDA-1; Tris(3,6-dioxaheptyl)amine
Molecular Weight (g/mol) 323.43
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.