Learn More
Medchemexpress LLC Tris(2-(3-tert-butylphenyl)-4-fluoropyridine)iridium(III) | 1311386-93-2 | MFCD30737992 | 98.0% | 877.07 g·mol-1 | C45H45F3IrN3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW03559950MG
Encompass_Preferred
Tris(2-(3-tert-butylphenyl)-4-fluoropyridine)iridium(III) is a cyclometalated iridium complex commonly used in research as a photocatalyst and emissive molecule. Its tris-chelated structure imparts strong photoluminescence and useful redox properties, making it suitable for visible-light photoredox reactions and optoelectronic studies.
  • Facilitates visible-light photoredox transformations.
  • Exhibits strong photoluminescence and long-lived excited states.
  • Provides reproducible redox behavior for catalytic studies.
  • Applicable to OLED and materials research for emissive layers.
  • Supplied as a stable solid suitable for low-temperature storage.
  • Available in small laboratory-scale quantities for screening and development.
Catalog No. 50-002-99562