Learn More
Medchemexpress LLC Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium | 1311386-93-2 | 877.07 g·mol⁻1 | C45H45F3IrN3 | 5 MG
SDP
Catalog No. 5000300179
Encompass_Preferred
Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium is an organometallic iridium(III) complex used as a photocatalyst and photoelectrochemical reagent for research. It demonstrates strong catalytic activity in visible-light-driven organic transformations and is applied in developing greener synthetic routes and photoelectrochemical studies.
  • Effective photocatalyst for visible-light-driven organic reactions.
  • Applicable to photoelectrochemical device and materials research.
  • Promotes efficient bond formation under mild conditions.
  • Molecular formula C45H45F3IrN3; molecular weight 877.07 g·mol⁻1.
  • Supplied for research use with standard handling and storage recommendations.
Catalog No. 50-003-00179 Supplier Medchemexpress LLC Supplier No. HYW0355995MG
Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium is an organometallic iridium(III) complex used as a photocatalyst and photoelectrochemical reagent for research. It demonstrates strong catalytic activity in visible-light-driven organic transformations and is applied in developing greener synthetic routes and photoelectrochemical studies.
  • Effective photocatalyst for visible-light-driven organic reactions.
  • Applicable to photoelectrochemical device and materials research.
  • Promotes efficient bond formation under mild conditions.
  • Molecular formula C45H45F3IrN3; molecular weight 877.07 g·mol⁻1.
  • Supplied for research use with standard handling and storage recommendations.