Learn More
Medchemexpress LLC Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium | 1311386-93-2 | MFCD30737992 | 98.0% | 877.07 | C45H45F3IrN3 | 25 MG
SDP
Catalog No. 5000303932
Encompass_Preferred
Ir[p-F(t-Bu)-ppy]3 is an iridium(III) organometallic complex used as a photoredox and photoluminescent reagent in research. It has molecular formula C45H45F3IrN3 and molecular weight 877.07. The compound should be protected from light and handled under inert atmosphere; in solution it is stable at -80°C for up to 6 months or -20°C for 1 month.
Catalog No. 50-003-03932 Supplier Medchemexpress LLC Supplier No. 25MGHYW035599
Ir[p-F(t-Bu)-ppy]3 is an iridium(III) organometallic complex used as a photoredox and photoluminescent reagent in research. It has molecular formula C45H45F3IrN3 and molecular weight 877.07. The compound should be protected from light and handled under inert atmosphere; in solution it is stable at -80°C for up to 6 months or -20°C for 1 month.