Learn More
Medchemexpress LLC Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium | 1311386-93-2 | MFCD30737992 | 98.0% | 877.07 | C45H45F3IrN3 | 250 MG
SDP
Catalog No. 5000303055
Encompass_Preferred
Ir[p-F(t-Bu)-ppy]3 is an organometallic iridium(III) complex used as a photocatalyst for visible-light-mediated organic synthesis and photoelectrochemical research. Supplied for laboratory use, it appears as a light-green solid with molecular formula C45H45F3IrN3 and molecular weight 877.07, and is handled under inert, light-protected conditions.
  • High photoredox catalytic activity for organic transformations.
  • Light-green solid suitable for handling in dry, inert conditions.
  • Recommended storage: protect from light and store under inert atmosphere.
  • Available in small vial quantities for lab-scale experiments.
  • Well-characterized composition for reproducible experimental results.
Catalog No. 50-003-03055 Supplier Medchemexpress LLC Supplier No. 250MGHYW035599
Ir[p-F(t-Bu)-ppy]3 is an organometallic iridium(III) complex used as a photocatalyst for visible-light-mediated organic synthesis and photoelectrochemical research. Supplied for laboratory use, it appears as a light-green solid with molecular formula C45H45F3IrN3 and molecular weight 877.07, and is handled under inert, light-protected conditions.
  • High photoredox catalytic activity for organic transformations.
  • Light-green solid suitable for handling in dry, inert conditions.
  • Recommended storage: protect from light and store under inert atmosphere.
  • Available in small vial quantities for lab-scale experiments.
  • Well-characterized composition for reproducible experimental results.