Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium | 1311386-93-2 | MFCD30737992 | 98.0% | 877.07 g·mol⁻¹ | C45H45F3IrN3 | 100 MG
SDP

Catalog No. 5000301548
Encompass_Preferred

Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium is an iridium-based photocatalyst used in visible-light mediated (photoredox) synthesis and biochemical assay applications. It is supplied as a pale-yellow to yellow-brown solid with high purity and specific storage requirements to maintain stability and performance.

  • Iridium-based photocatalyst for photoredox reactions.
  • High purity (~98.0%) suitable for research applications.
  • Molecular formula C45H45F3IrN3 and molecular weight 877.07 g·mol⁻¹.
  • CAS number 1311386-93-2 for unambiguous identification.
  • Store at 4°C, protect from light, and keep under inert atmosphere; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-01548 Supplier Medchemexpress LLC Supplier No. HYW035599100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium is an iridium-based photocatalyst used in visible-light mediated (photoredox) synthesis and biochemical assay applications. It is supplied as a pale-yellow to yellow-brown solid with high purity and specific storage requirements to maintain stability and performance.

  • Iridium-based photocatalyst for photoredox reactions.
  • High purity (~98.0%) suitable for research applications.
  • Molecular formula C45H45F3IrN3 and molecular weight 877.07 g·mol⁻¹.
  • CAS number 1311386-93-2 for unambiguous identification.
  • Store at 4°C, protect from light, and keep under inert atmosphere; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.