Learn More
Medchemexpress LLC Tris(2-(3-tert-butylphenyl)-4-tert-butylpyridine)iridium | 1311386-93-2 | MFCD30737992 | 98.0% | 880.1 g/mol | C45H48F3IrN3 | 10 MG
SDP
Catalog No. 5000302071
Encompass_Preferred
Ir[p-F(t-Bu)-ppy]3 is a cyclometalated iridium(III) complex used in research as a photoredox catalyst and as a phosphorescent emitter for optoelectronic studies. It is applied in photocatalytic organic transformations and in the development and testing of OLED materials and devices.
  • Serves as an efficient photoredox catalyst for organic synthesis.
  • Functions as a phosphorescent emitter for optoelectronic and OLED research.
  • High purity (98.0%).
  • Molecular formula C45H48F3IrN3; molecular weight 880.1 g/mol.
  • Offered in small research pack sizes suitable for laboratory use.
Catalog No. 50-003-02071 Supplier Medchemexpress LLC Supplier No. HYW03559910MG
Ir[p-F(t-Bu)-ppy]3 is a cyclometalated iridium(III) complex used in research as a photoredox catalyst and as a phosphorescent emitter for optoelectronic studies. It is applied in photocatalytic organic transformations and in the development and testing of OLED materials and devices.
  • Serves as an efficient photoredox catalyst for organic synthesis.
  • Functions as a phosphorescent emitter for optoelectronic and OLED research.
  • High purity (98.0%).
  • Molecular formula C45H48F3IrN3; molecular weight 880.1 g/mol.
  • Offered in small research pack sizes suitable for laboratory use.