Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(2,4,6-Trimethylphenyl)Phosphine

Catalog No. 3950805G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-508-05G 5 g
39-508-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-508-05G Supplier Sigma Aldrich Supplier No. 3950805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(2,4,6-Trimethylphenyl)Phosphine

Specifications

CAS 23897-15-6
Melting Point 185°C to 188°C
Molecular Formula C27H33P
Linear Formula [(CH3)3C6H2]3P
MDL Number MFCD00014912
Quantity 5 g
Synonym Trimesitylphosphine
Molecular Weight (g/mol) 388.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.