Learn More
Medchemexpress LLC HY-136177 100mg Medchemexpress, Tris(2,4-di-tert-butylphenyl)phosphate CAS:95906-11-9 Purity:>98%
SDP
Catalog No. 502030403
Encompass_Preferred
Medchemexpress, HY-136177 100mg Tris(2,4-di-tert-butylphenyl)phosphate CAS:95906-11-9 Tris(2,4-di-tert-butylphenyl)phosphate is an active compound from the leaves of Vitex negundo L. shows anti-inflammatory activity with evidence of inhibition for secretory Phospholipase A2 (sPLA2) through molecular docking. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0403 Supplier Medchemexpress LLC Supplier No. HY136177100MG
Medchemexpress, HY-136177 100mg Tris(2,4-di-tert-butylphenyl)phosphate CAS:95906-11-9 Tris(2,4-di-tert-butylphenyl)phosphate is an active compound from the leaves of Vitex negundo L. shows anti-inflammatory activity with evidence of inhibition for secretory Phospholipase A2 (sPLA2) through molecular docking. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.