Learn More
Medchemexpress LLC Tris(2,4-di-tert-butylphenyl)phosphate | 95906-11-9 | 99.8% | 662.92 | 25 MG
SDP
Catalog No. 5000348879
Encompass_Preferred
Tris(2,4-di-tert-butylphenyl)phosphate is an active compound derived from the leaves of Vitex negundo L. It exhibits anti-inflammatory activity by inhibiting secretory Phospholipase A2 (sPLA2) through molecular docking.
  • Active compound from leaves of Vitex negundo L.
  • Demonstrates anti-inflammatory activity.
  • Inhibits secretory Phospholipase A2 (sPLA2) via molecular docking.
  • Shows significant anti-inflammatory activity in carrageenan-induced paw edema model at 50 mg/kg and 70 mg/kg doses.
Catalog No. 50-003-48879 Supplier Medchemexpress LLC Supplier No. 25MGHY136177R
Tris(2,4-di-tert-butylphenyl)phosphate is an active compound derived from the leaves of Vitex negundo L. It exhibits anti-inflammatory activity by inhibiting secretory Phospholipase A2 (sPLA2) through molecular docking.
  • Active compound from leaves of Vitex negundo L.
  • Demonstrates anti-inflammatory activity.
  • Inhibits secretory Phospholipase A2 (sPLA2) via molecular docking.
  • Shows significant anti-inflammatory activity in carrageenan-induced paw edema model at 50 mg/kg and 70 mg/kg doses.