Learn More
Medchemexpress LLC Tris(2,4-di-tert-butylphenyl)phosphate | 95906-11-9 | 99.8% | 662.92 | 50 MG
SDP
Catalog No. 5000351612
Encompass_Preferred
Tris(2,4-di-tert-butylphenyl)phosphate is an active compound derived from the leaves of Vitex negundo L. This compound demonstrates anti-inflammatory activity, primarily by inhibiting secretory Phospholipase A2 (sPLA2) through molecular docking mechanisms. It has shown significant anti-inflammatory effects in in vivo studies.
  • Inhibits secretory Phospholipase A2 (sPLA2)
  • Derived from Vitex negundo L.
  • Demonstrates anti-inflammatory activity
  • Significant anti-inflammatory effects in carrageenan-induced paw edema model
Catalog No. 50-003-51612 Supplier Medchemexpress LLC Supplier No. HY13617750MG
Tris(2,4-di-tert-butylphenyl)phosphate is an active compound derived from the leaves of Vitex negundo L. This compound demonstrates anti-inflammatory activity, primarily by inhibiting secretory Phospholipase A2 (sPLA2) through molecular docking mechanisms. It has shown significant anti-inflammatory effects in in vivo studies.
  • Inhibits secretory Phospholipase A2 (sPLA2)
  • Derived from Vitex negundo L.
  • Demonstrates anti-inflammatory activity
  • Significant anti-inflammatory effects in carrageenan-induced paw edema model