Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tris(2,4-di-tert-butylphenyl)phosphate | 95906-11-9 | MFCD29918884 | 99.7% | C42H63O4P | 10MG
SDP

Catalog No. 5000259904
Encompass_Preferred

Tris(2,4-di-tert-butylphenyl)phosphate is an organophosphate analytical standard (CAS 95906-11-9) supplied as a high-purity reference material for research and analytical applications. It is used for calibration, assay validation, and polymer stabilization studies, and is available in small laboratory pack sizes.

  • High purity (99.7%).
  • Molecular formula: C42H63O4P.
  • Molecular weight: 662.92 g/mol.
  • Intended use: analytical reference, calibration, and method development.
  • Available in small pack sizes suitable for laboratory use.

Catalog No. 50-002-59904 Supplier Medchemexpress LLC Supplier No. HY136177R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris(2,4-di-tert-butylphenyl)phosphate is an organophosphate analytical standard (CAS 95906-11-9) supplied as a high-purity reference material for research and analytical applications. It is used for calibration, assay validation, and polymer stabilization studies, and is available in small laboratory pack sizes.

  • High purity (99.7%).
  • Molecular formula: C42H63O4P.
  • Molecular weight: 662.92 g/mol.
  • Intended use: analytical reference, calibration, and method development.
  • Available in small pack sizes suitable for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.