Learn More
Medchemexpress LLC Tris(2,4-di-tert-butylphenyl)phosphate | 95906-11-9 | MFCD29918884 | 99.8% | 662.92 | 250 MG
SDP
Catalog No. 5000351110
Encompass_Preferred
Tris(2,4-di-tert-butylphenyl)phosphate is an active compound derived from the leaves of Vitex negundo L. It exhibits anti-inflammatory activity, demonstrated by its inhibition of secretory Phospholipase A2 (sPLA2) through molecular docking.
  • Derived from the leaves of Vitex negundo L.
  • Exhibits anti-inflammatory activity.
  • Inhibits secretory Phospholipase A2 (sPLA2) through molecular docking.
  • Reduces paw edema volume in a carrageenan-induced paw edema model.
Catalog No. 50-003-51110 Supplier Medchemexpress LLC Supplier No. 250MGHY136177
Tris(2,4-di-tert-butylphenyl)phosphate is an active compound derived from the leaves of Vitex negundo L. It exhibits anti-inflammatory activity, demonstrated by its inhibition of secretory Phospholipase A2 (sPLA2) through molecular docking.
  • Derived from the leaves of Vitex negundo L.
  • Exhibits anti-inflammatory activity.
  • Inhibits secretory Phospholipase A2 (sPLA2) through molecular docking.
  • Reduces paw edema volume in a carrageenan-induced paw edema model.