Learn More
Medchemexpress LLC Tris(2,4-di-tert-butylphenyl)phosphate | 95906-11-9 | MFCD29918884 | 99.8% | 662.92 | 500 MG
SDP
Catalog No. 5000350910
Encompass_Preferred
Tris(2,4-di-tert-butylphenyl)phosphate is an active compound derived from the leaves of Vitex negundo L. It demonstrates anti-inflammatory activity by inhibiting secretory Phospholipase A2 (sPLA2) through molecular docking. In vivo studies show that at doses of 50 mg/kg and 70 mg/kg, it significantly reduces paw edema volume in a carrageenan-induced paw edema model.
  • Active compound from Vitex negundo L.
  • Demonstrates anti-inflammatory activity
  • Inhibits secretory Phospholipase A2 (sPLA2)
  • Effective in reducing paw edema volume in vivo
Catalog No. 50-003-50910 Supplier Medchemexpress LLC Supplier No. HY136177500MG
Tris(2,4-di-tert-butylphenyl)phosphate is an active compound derived from the leaves of Vitex negundo L. It demonstrates anti-inflammatory activity by inhibiting secretory Phospholipase A2 (sPLA2) through molecular docking. In vivo studies show that at doses of 50 mg/kg and 70 mg/kg, it significantly reduces paw edema volume in a carrageenan-induced paw edema model.
  • Active compound from Vitex negundo L.
  • Demonstrates anti-inflammatory activity
  • Inhibits secretory Phospholipase A2 (sPLA2)
  • Effective in reducing paw edema volume in vivo