Learn More
Medchemexpress LLC Tris(2,4-di-tert-butylphenyl)phosphate (Standard) | 95906-11-9 | MFCD29918884 | 99.73% | 662.92 | 50 MG
SDP
Catalog No. 5000350911
Encompass_Preferred
Tris(2,4-di-tert-butylphenyl)phosphate (Standard) is the analytical standard of Tris(2,4-di-tert-butylphenyl)phosphate, intended for research and analytical applications. It is an active compound from the leaves of Vitex negundo L., demonstrating anti-inflammatory activity through the inhibition of secretory Phospholipase A2 (sPLA2) via molecular docking.
  • Analytical standard for research and analytical applications.
  • Derived from Vitex negundo L., showing anti-inflammatory activity.
  • Inhibits secretory Phospholipase A2 (sPLA2).
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-003-50911 Supplier Medchemexpress LLC Supplier No. HY136177R50MG
Tris(2,4-di-tert-butylphenyl)phosphate (Standard) is the analytical standard of Tris(2,4-di-tert-butylphenyl)phosphate, intended for research and analytical applications. It is an active compound from the leaves of Vitex negundo L., demonstrating anti-inflammatory activity through the inhibition of secretory Phospholipase A2 (sPLA2) via molecular docking.
  • Analytical standard for research and analytical applications.
  • Derived from Vitex negundo L., showing anti-inflammatory activity.
  • Inhibits secretory Phospholipase A2 (sPLA2).
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.