Learn More
Medchemexpress LLC Tris(2-(4-fluorophenyl)pyridine)iridium | 370878-69-6 | 98.0% | 708.75 | C33H21F3IrN3 | 50 MG
SDP
Catalog No. 5000299858
Encompass_Preferred
Ir(p-F-ppy)3 (Tris(2-(4-fluorophenyl)pyridine)iridium) is an organometallic iridium(III) complex used in photophysical and optoelectronic research. Supplied as a light yellow solid with high purity, it is suitable for studies of photoluminescence, electroluminescence, and photocatalysis.
  • High purity (98.0%) suitable for research applications.
  • Solid, light yellow to yellow appearance for easy handling.
  • Useful as a phosphorescent emitter in OLED and photoluminescence studies.
  • Suitable for photocatalysis and photochemical investigations.
  • Stable when stored protected from light; solid storage at 4°C under inert atmosphere advised.
  • Molecular formula C33H21F3IrN3; molecular weight 708.75.
Catalog No. 50-002-99858 Supplier Medchemexpress LLC Supplier No. HYW03404150MG
Ir(p-F-ppy)3 (Tris(2-(4-fluorophenyl)pyridine)iridium) is an organometallic iridium(III) complex used in photophysical and optoelectronic research. Supplied as a light yellow solid with high purity, it is suitable for studies of photoluminescence, electroluminescence, and photocatalysis.
  • High purity (98.0%) suitable for research applications.
  • Solid, light yellow to yellow appearance for easy handling.
  • Useful as a phosphorescent emitter in OLED and photoluminescence studies.
  • Suitable for photocatalysis and photochemical investigations.
  • Stable when stored protected from light; solid storage at 4°C under inert atmosphere advised.
  • Molecular formula C33H21F3IrN3; molecular weight 708.75.