Learn More
Medchemexpress LLC Tris(2-(4-fluorophenyl)pyridine)iridium | 370878-69-6 | 98.0% | 708.75 g·mol⁻¹ | C33H21F3IrN3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW03404110MG
Encompass_Preferred
Ir(p-F-ppy)3 (Tris(2-(4-fluorophenyl)pyridine)iridium) is a cyclometalated iridium(III) complex used as a phosphorescent emitter and dopant in optoelectronic research. It is supplied as a light yellow to yellow solid with high purity for photophysical studies, organic light-emitting diode development, and related photocatalysis research.
  • High purity: 98.0% as supplied.
  • Light yellow to yellow solid appearance.
  • Used as a phosphorescent emitter and dopant in OLEDs and photophysical studies.
  • Multiple small pack sizes available for research use (5-500 mg).
  • Molecular weight 708.75 g·mol⁻¹ and formula C33H21F3IrN3.
Catalog No. 50-003-01777