Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tris(2-(4-fluorophenyl)pyridine)iridium | 370878-69-6 | MFCD30830351 | 98.0% | 708.75 g·mol⁻¹ | C33H21F3IrN3 | 100 MG
SDP

Catalog No. 5000300097
Encompass_Preferred

Ir(p-F-ppy)3 is an iridium(III) coordination complex used as a visible-light photocatalyst in photoredox organic synthesis and optoelectronic research. It exhibits strong photoluminescence and efficient electron transfer, enabling improved reaction selectivity and device performance.

  • High purity (98.0%) confirmed by NMR.
  • Molecular formula C33H21F3IrN3.
  • Molecular weight 708.75 g·mol⁻¹.
  • Effective visible-light photocatalyst for photoredox transformations.
  • Suitable for photophysical studies and optoelectronic device research.

Catalog No. 50-003-00097 Supplier Medchemexpress LLC Supplier No. HYW034041100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ir(p-F-ppy)3 is an iridium(III) coordination complex used as a visible-light photocatalyst in photoredox organic synthesis and optoelectronic research. It exhibits strong photoluminescence and efficient electron transfer, enabling improved reaction selectivity and device performance.

  • High purity (98.0%) confirmed by NMR.
  • Molecular formula C33H21F3IrN3.
  • Molecular weight 708.75 g·mol⁻¹.
  • Effective visible-light photocatalyst for photoredox transformations.
  • Suitable for photophysical studies and optoelectronic device research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.