Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2,6-dimethoxyphenyl)phosphine 97.0+%, TCI America™
SDP

Catalog No. T161425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T161425G 25 g
T1614-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T161425G Supplier TCI America Supplier No. T161425G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Tris(2,6-dimethoxyphenyl)phosphine
CAS 85417-41-0
Melting Point 155°C
Color White-Yellow
Molecular Formula C24H27O6P
Quantity 25 g
MDL Number MFCD00192558
Synonym tris 2,6-dimethoxyphenyl phosphine, tris 2,6-dimethoxyphenyl phosphane, tdmpp, phosphine, tris 2,6-dimethoxyphenyl, tris 2,6-dimethoxyphenly phosphine, pubchem6450, acmc-209q5v, 2,6-dmpp, cmlwfcuaxgsmbb-uhfffaoysa, tri 2,6-dimethoxyphenyl phosphine
InChI Key CMLWFCUAXGSMBB-UHFFFAOYSA-N
SMILES COC1=C(C(=CC=C1)OC)P(C2=C(C=CC=C2OC)OC)C3=C(C=CC=C3OC)OC
IUPAC Name tris(2,6-dimethoxyphenyl)phosphane
Molecular Weight (g/mol) 442.448
PubChem CID 2734597
Formula Weight 442.45
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.