Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris[2-(acryloyloxy)ethyl] isocyanurate

Catalog No. 407534250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-753-4250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-753-4250G Supplier Sigma Aldrich Supplier No. 407534250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris[2-(acryloyloxy)ethyl] isocyanurate

Specifications

CAS 40220-08-4
Melting Point 52°C to 53°C
Molecular Formula C18H21N3O9
Linear Formula C18H21N3O9
Quantity 250 g
MDL Number MFCD00080669
Synonym Tri(2-hydroxyethyl)isocyanurate triacrylate; Tris(2-hydroxyethyl) isocyanurate triacrylate; Tris(2-hydroxyethyl)isocyanuric acid triacrylate; Tris[2-(acryloyloxy)ethyl]
Molecular Weight (g/mol) 423.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.