Learn More
eMolecules​ 51805-45-9 | Tris(2-carboxyethyl)phosphine hydrochloride | Combi-Blocks | MFCD00145469 | 286.640 | C9H16ClO6P | 97.000 | Cl.OC(=O)CCP(CCC(O)=O)CCC(O)=O | 25g | 232306938
Catalog No. NC2020604 Shop All eMolecules Inc. Products
Encompass_Preferred
Tris(2-carboxyethyl)phosphine hydrochloride | Combi-Blocks | 51805-45-9 | MFCD00145469 | 286.640 | C9H16ClO6P | 97.000 | Cl.OC(=O)CCP(CCC(O)=O)CCC(O)=O | 25g | 232306938
Catalog No. NC2020604 Supplier eMolecules​ Supplier No. 232306938
Tris(2-carboxyethyl)phosphine hydrochloride | Combi-Blocks | 51805-45-9 | MFCD00145469 | 286.640 | C9H16ClO6P | 97.000 | Cl.OC(=O)CCP(CCC(O)=O)CCC(O)=O | 25g | 232306938