Learn More
Strem, An Ascensus Company CAS# 51805-45-9. 1g. Tris(2-carboxyethyl)phosphine, hydrochloride, 99% TCEP. MFCD00145469
SDP
Catalog No. 5090110492
Encompass
CAS# 51805-45-9. 1g. Tris(2-carboxyethyl)phosphine, hydrochloride, 99% TCEP. MFCD00145469. Molecular Weight: 286.65. Molecular Formula: (HOOCCH2CH2)3PH+Clae. Color/form: white xtl. Strem# 15-7400. http://www.strem.com/catalog/v/15-7400/
Catalog No. 50-901-10492 Supplier Strem, An Ascensus Company Supplier No. 1574001G
CAS# 51805-45-9. 1g. Tris(2-carboxyethyl)phosphine, hydrochloride, 99% TCEP. MFCD00145469. Molecular Weight: 286.65. Molecular Formula: (HOOCCH2CH2)3PH+Clae. Color/form: white xtl. Strem# 15-7400. http://www.strem.com/catalog/v/15-7400/