Learn More
Crescent Chemical Co Inc TRIS-(2-CARBOXYETHYL)PHOSPHINE
SDP
Catalog No. 50248213
Encompass_Preferred
Tris-(2-carboxyethyl)phosphine hydrochloride Water-soluble and odorless reagent for selective and fast reduction of disulfides. Does not react with other functional groups of proteins. Unreactive towards many common alkylating reagents, so reductions have been carried out simultaneously wit
Catalog No. 50-248-213 Supplier Crescent Chemical Co Inc Supplier No. SE36970.01
Tris-(2-carboxyethyl)phosphine hydrochloride Water-soluble and odorless reagent for selective and fast reduction of disulfides. Does not react with other functional groups of proteins. Unreactive towards many common alkylating reagents, so reductions have been carried out simultaneously wit