Learn More
Medchemexpress LLC Tris(2-carboxyethyl)phosphine-d12 hydrochloride | 1189959-10-1 | 98.0% | 298.72 g/mol | C9H4D12ClO6P | 1 MG
SDP
Catalog No. 5000284534
Encompass_Preferred
TCEP-d12 hydrochloride is the deuterated hydrochloride salt of tris(2-carboxyethyl)phosphine, used as a non-thiol reducing agent for disulfide bond reduction and as an isotopic standard in analytical workflows.
  • Deuterated hydrochloride isotopologue for mass spectrometry applications.
  • Non-thiol reducing agent for efficient disulfide bond reduction in proteins and peptides.
  • High purity suitable for analytical and research use (verify lot CoA for exact purity).
  • Provided in a small laboratory pack for trace-level or standard preparation.
  • Stable and water-soluble hydrochloride salt for convenient handling.
Catalog No. 50-002-84534 Supplier Medchemexpress LLC Supplier No. HYW7006211MG
TCEP-d12 hydrochloride is the deuterated hydrochloride salt of tris(2-carboxyethyl)phosphine, used as a non-thiol reducing agent for disulfide bond reduction and as an isotopic standard in analytical workflows.
  • Deuterated hydrochloride isotopologue for mass spectrometry applications.
  • Non-thiol reducing agent for efficient disulfide bond reduction in proteins and peptides.
  • High purity suitable for analytical and research use (verify lot CoA for exact purity).
  • Provided in a small laboratory pack for trace-level or standard preparation.
  • Stable and water-soluble hydrochloride salt for convenient handling.