Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-carboxyethyl)phosphine hydrochloride Solution 0.5M, MilliporeSigma™ Supelco™

Catalog No. 111014418 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4418 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4418 Supplier MilliporeSigma Supelco Supplier No. 64654710X1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

It belongs to the trialkylphosphine class.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51805-45-9
Density 1.041 g/mL (at 25°C)
Concentration 0.5 M
Linear Formula C9H15O6 P · HCl
MDL Number MFCD00145469
pH 7.0 (aq. soln.; pH was adjusted with ammonium hydroxide)
Synonym TCEP
Type Organic Standard
Quantity 10 x 1 mL
Formula Weight 286.65
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.