Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tris(2-chloroethyl) phosphate | 115-96-8 | MFCD00000967 | 97.1% | 285.49 | C6H12Cl3O4P | 50 MG
SDP

Catalog No. 5000240285
Encompass_Preferred

Tris(2-chloroethyl) phosphate is an organophosphate compound used as a flame retardant, plasticizer, and laboratory reagent. It is supplied as a liquid reagent with documented chemical properties, typical purity, and storage recommendations for research and industrial use.

  • CAS number 115-96-8.
  • Molecular formula C6H12Cl3O4P.
  • Molecular weight 285.49 g·mol⁻¹.
  • Purity about 97.1%.
  • Physical state liquid.
  • Recommended storage: room temperature for bulk material; in solvent store at -80 °C (2 years) or -20 °C (1 year).
  • Available in research pack sizes from 5 mg to 500 mg.

Catalog No. 50-002-40285 Supplier Medchemexpress LLC Supplier No. HY10785950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris(2-chloroethyl) phosphate is an organophosphate compound used as a flame retardant, plasticizer, and laboratory reagent. It is supplied as a liquid reagent with documented chemical properties, typical purity, and storage recommendations for research and industrial use.

  • CAS number 115-96-8.
  • Molecular formula C6H12Cl3O4P.
  • Molecular weight 285.49 g·mol⁻¹.
  • Purity about 97.1%.
  • Physical state liquid.
  • Recommended storage: room temperature for bulk material; in solvent store at -80 °C (2 years) or -20 °C (1 year).
  • Available in research pack sizes from 5 mg to 500 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.