Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tris(2-chloroethyl) phosphate | 115-96-8 | MFCD00000967 | 97.1% | 285.49 g/mol | C6H12Cl3O4P | 100 MG
SDP

Catalog No. 5000240825
Encompass_Preferred

Tris(2-chloroethyl) phosphate is an organophosphate triester used as a research reagent and industrial flame retardant. Supplied as a colorless to light yellow liquid with typical high purity, it is used in polymer formulations, analytical standards, and laboratory studies for toxicology and materials research.

  • CAS number 115-96-8.
  • Molecular formula C6H12Cl3O4P.
  • Molecular weight 285.49 g/mol.
  • Typical purity 97.1%.
  • Physical form colorless to light yellow liquid.
  • Applications include flame retardant, plasticizer, and laboratory reagent.
  • Storage: store at room temperature; in solvent, keep at -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-002-40825 Supplier Medchemexpress LLC Supplier No. HY107859100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris(2-chloroethyl) phosphate is an organophosphate triester used as a research reagent and industrial flame retardant. Supplied as a colorless to light yellow liquid with typical high purity, it is used in polymer formulations, analytical standards, and laboratory studies for toxicology and materials research.

  • CAS number 115-96-8.
  • Molecular formula C6H12Cl3O4P.
  • Molecular weight 285.49 g/mol.
  • Typical purity 97.1%.
  • Physical form colorless to light yellow liquid.
  • Applications include flame retardant, plasticizer, and laboratory reagent.
  • Storage: store at room temperature; in solvent, keep at -80°C for 2 years or -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.