Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tris(2-chloroethyl) phosphate | 115-96-8 | MFCD00000967 | 97.1% | 285.49 g/mol | C6H12Cl3O4P | 25 MG
SDP

Catalog No. 5000242314
Encompass_Preferred

Tris(2-chloroethyl) phosphate is an organophosphate triester used as a research reagent, flame retardant, plasticizer, and viscosity regulator in polymer and materials research. It is supplied as a colorless to light-yellow liquid with a reported molecular weight of 285.49 g/mol and typical purity around 97%.

  • Organophosphate flame retardant and plasticizer suitable for research applications.
  • Colorless to light yellow liquid appearance.
  • Molecular weight 285.49 g/mol.
  • Typical purity approximately 97.1%.
  • Available in small quantities tailored for laboratory use.
  • Storage stability at room temperature for up to 3 years; in solvent store at -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-002-42314 Supplier Medchemexpress LLC Supplier No. HY10785925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris(2-chloroethyl) phosphate is an organophosphate triester used as a research reagent, flame retardant, plasticizer, and viscosity regulator in polymer and materials research. It is supplied as a colorless to light-yellow liquid with a reported molecular weight of 285.49 g/mol and typical purity around 97%.

  • Organophosphate flame retardant and plasticizer suitable for research applications.
  • Colorless to light yellow liquid appearance.
  • Molecular weight 285.49 g/mol.
  • Typical purity approximately 97.1%.
  • Available in small quantities tailored for laboratory use.
  • Storage stability at room temperature for up to 3 years; in solvent store at -80°C for 2 years or -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.