Learn More
Medchemexpress LLC Tris(2-chloroethyl) phosphate | 115-96-8 | MFCD00000967 | 97.1% | 285.49 g/mol | C6H12Cl3O4P | 5 MG
SDP
Catalog No. 5000244507
Encompass_Preferred
Tris(2-chloroethyl) phosphate is an organophosphate triester supplied as a research reagent. It is used as a flame retardant and plasticizer and is provided as a colorless to light yellow liquid with defined storage and stability. Manufacturer documentation includes a datasheet, certificate of analysis, and safety data sheet for handling and analytical details.
  • High purity verified by GC.
  • Functions as a flame retardant and plasticizer.
  • Colorless to light yellow liquid appearance.
  • Stable at room temperature with extended shelf life.
  • Datasheet, COA, and SDS available for analytical and safety data.
Catalog No. 50-002-44507 Supplier Medchemexpress LLC Supplier No. HY1078595MG
Tris(2-chloroethyl) phosphate is an organophosphate triester supplied as a research reagent. It is used as a flame retardant and plasticizer and is provided as a colorless to light yellow liquid with defined storage and stability. Manufacturer documentation includes a datasheet, certificate of analysis, and safety data sheet for handling and analytical details.
  • High purity verified by GC.
  • Functions as a flame retardant and plasticizer.
  • Colorless to light yellow liquid appearance.
  • Stable at room temperature with extended shelf life.
  • Datasheet, COA, and SDS available for analytical and safety data.