Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tris(2-chloroethyl) phosphate-d12 | 1276500-47-0 | MFCD20257118 | 99.2% | 297.56 | C6D12Cl3O4P | 10 MG
SDP

Catalog No. 5000242312
Encompass_Preferred

Tris(2-chloroethyl)phosphate-d12 is the deuterium-labeled analogue of tris(2-chloroethyl) phosphate used as a stable isotope tracer or internal standard in analytical and research workflows. It is supplied as a high-purity, isotopically labeled reagent intended for laboratory research use only.

  • Deuterium-labeled internal standard for analytical assays.
  • High purity (99.2%).
  • Chemical formula C6D12Cl3O4P; molecular weight 297.56.
  • Liquid, colorless to light yellow.
  • CAS number 1276500-47-0 for unambiguous identification.
  • Store at -20°C for long-term storage; 4°C for short-term storage in pure form.
  • For research use only; not for clinical or patient use.

Catalog No. 50-002-42312 Supplier Medchemexpress LLC Supplier No. HY107859S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris(2-chloroethyl)phosphate-d12 is the deuterium-labeled analogue of tris(2-chloroethyl) phosphate used as a stable isotope tracer or internal standard in analytical and research workflows. It is supplied as a high-purity, isotopically labeled reagent intended for laboratory research use only.

  • Deuterium-labeled internal standard for analytical assays.
  • High purity (99.2%).
  • Chemical formula C6D12Cl3O4P; molecular weight 297.56.
  • Liquid, colorless to light yellow.
  • CAS number 1276500-47-0 for unambiguous identification.
  • Store at -20°C for long-term storage; 4°C for short-term storage in pure form.
  • For research use only; not for clinical or patient use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.