Learn More
Strem, An Ascensus Company CAS# 4023-53-4. 25g. Tris(2-cyanoethyl)phosphine, min. 99%. MFCD00013828
SDP
Catalog No. 502045261
Encompass
CAS# 4023-53-4. 25g. Tris(2-cyanoethyl)phosphine, min. 99%. MFCD00013828. Molecular Weight: 193.19. Molecular Formula: P(CH2CH2CN)3. Color/form: white xtl. Strem# 15-7680. http://www.strem.com/catalog/v/15-7680/
Catalog No. 50-204-5261 Supplier Strem, An Ascensus Company Supplier No. 15768025G
CAS# 4023-53-4. 25g. Tris(2-cyanoethyl)phosphine, min. 99%. MFCD00013828. Molecular Weight: 193.19. Molecular Formula: P(CH2CH2CN)3. Color/form: white xtl. Strem# 15-7680. http://www.strem.com/catalog/v/15-7680/