Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-cyanoethyl)phosphine, 94%

Catalog No. AA3016509
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3016509 10 g
AA3016503 1 g
AA3016518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3016509 Supplier Thermo Scientific Chemicals Supplier No. 03016509
Only null left
Add to Cart

CAS: 4023-53-4 | C9H12N3P | 193.19 g/mol

Tris(2-cyanoethyl)phosphine is used to produce 3,3',3″-phosphanetriyl-tri-propionic acid and hydrochloride by heating. It will need reagent conc. aq. HCl with reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4023-53-4
Molecular Formula C9H12N3P
Molecular Weight (g/mol) 193.19
MDL Number MFCD00013828
InChI Key CHZAMJVESILJGH-UHFFFAOYSA-N
Synonym tris 2-cyanoethyl phosphine, 3,3',3-phosphinetriyltripropanenitrile, tris-2-cyanoethyl phosphine, tris-2-cyanoethyl-phosphine, propanenitrile, 3,3',3-phosphinidynetris, acmc-20ajc3, p ch2ch2cn 3, 3-bis 2-cyanoethyl phosphanyl propanenitrile, propanenitrile,3',3-phosphinidynetris, tris 2-cyanoethyl phosphine 10g
PubChem CID 77639
IUPAC Name 3-[bis(2-cyanoethyl)phosphanyl]propanenitrile
SMILES C(CP(CCC#N)CCC#N)C#N

Specifications

Melting Point 97°C to 98°C
Boiling Point 235°C (0.9 mmHg)
Quantity 10 g
UN Number UN3464
Sensitivity Air and moisture sensitive
Solubility Information Soluble in DMSO,and DMF. Insoluble in water.
Formula Weight 193.19
Percent Purity 94%
Physical Form Crystalline
Chemical Name or Material Tris(2-cyanoethyl)phosphine
Hazard Category H302-H315-H319-H331-H335-H350
Hazard Statement GHS H Statement
H300-H330-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
EINECSNumber 223-687-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.