Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris[2-(dimethylamino)ethyl]amine

Catalog No. 7231421ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-314-21ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-314-21ML Supplier Sigma Aldrich Supplier No. 7231421ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris[2-(dimethylamino)ethyl]amine (Me6TREN) is an aliphatic tertiary amine that forms a quadridentate ligand. It is majorly used in atom transfer radical polymerization (ATRP). It is a hexamethyl derivative of (2-aminoethyl)amine, also known as trenMe. It can be prepared by a modification of Ristempart's synthesis.Me6TREN forms a complex with copper compounds which can potentially be used as ligands in metalloenzyme redox reactions.

Specifications

CAS 33527-91-2
Density 0.862 g/mL (at 25°C)
Molecular Formula C12H30N4
Linear Formula C12H30N4
Refractive Index n20/D 1.455
MDL Number MFCD00015607
Quantity 1 mL
Synonym Me6TREN
Molecular Weight (g/mol) 230.39
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.