Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(2-ethylhexyl) phosphate

Catalog No. 28992225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-992-225ML 25 mL
28-992-2500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-992-225ML Supplier Sigma Aldrich Supplier No. 28992225ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(2-ethylhexyl) phosphate exhibits glucocorticoid receptor (GR) antagonistic activity.

Specifications

CAS 78-42-2
Density 0.92 g/mL (at 20°C (literature))
Boiling Point 215°C (4 mmHg)
Molecular Formula C24H51O4P
Refractive Index n20/D 1.444 (literature)
Linear Formula [CH3(CH2)3 CH(C2H5)CH2O]3 P(O)
MDL Number MFCD00009491
Quantity 25 mL
Synonym Trioctyl phosphate
Molecular Weight (g/mol) 434.63
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.