Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tris(2-Ethylhexyl)phosphate, 99.0%, MP Biomedicals™

Catalog No. ICN15799125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15799125 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15799125 Supplier MP Biomedicals, Inc Supplier No. 0215799125
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78-42-2
Molecular Formula C24H51O4P
Molecular Weight (g/mol) 434.64
MDL Number MFCD00009491
InChI Key GTVWRXDRKAHEAD-UHFFFAOYNA-N
Synonym tris 2-ethylhexyl phosphate, disflamoll tof, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
PubChem CID 6537
IUPAC Name tris(2-ethylhexyl) phosphate
SMILES CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC

Specifications

Color Colorless
Density 0.926 at 20°C
Assay Percent Range 99%
Quantity 25 mL
Formula Weight 434.63
Physical Form Liquid
Chemical Name or Material tris(2-Ethylhexyl)phosphate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.