Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-methylallyl)amine 99%, Thermo Scientific™

Catalog No. AC171861000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171861000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171861000 Supplier Thermo Scientific Chemicals Supplier No. 171861000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6321-40-0
Molecular Formula C12H21N
Molecular Weight (g/mol) 179.31
MDL Number MFCD00008596
InChI Key FXBJYRVIFGLPBC-UHFFFAOYSA-N
Synonym tris 2-methylallyl amine, trimethallylamine, ch2=c ch3 ch2 3n, 2-methyl-n,n-bis 2-methylprop-2-enyl prop-2-en-1-amine, tris 2-methylprop-2-en-1-yl amine, tris-2-methyl-allyl-amine, fxbjyrvifglpbc-uhfffaoysa, 2-methyl-n,n-bis 2-methylprop-2-en-1-yl prop-2-en-1-amine, 2-methyl-n,n-bis 2-methylallyl prop-2-en-1-amine
PubChem CID 80605
IUPAC Name tris(2-methylprop-2-en-1-yl)amine
SMILES CC(=C)CN(CC(C)=C)CC(C)=C

Specifications

Density 0.79
Linear Formula [H2C=C(CH3)CH2]3N
Quantity 100 g
Beilstein 04, III, 462
Formula Weight 179.3
Chemical Name or Material Tris(2-methylallyl)amine, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.