Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris[2-phenylpyridinato-C2,N]iridium(III)

Catalog No. 688096250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-809-6250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-809-6250MG Supplier Sigma Aldrich Supplier No. 688096250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris-(2-phenylpyridine) iridium [Ir(ppy)3] is a heavy metal complex. [Ir(ppy)3] is the most frequently used precursor molecule for the synthesis of electro-phosphorescent materials, which are then used in organic light emitting diodes (OLEDs). It provides green-color emission and high phosphorescence quantum yield close to unity.Put to elegant use by Stephenson for the reduction of alkyl, alkenyl and aryl iodides in the presence of visible light. Intramolecular reductive cyclizations were also achieved with this catalyst.Engaging unactivated alkyl, alkenyl and aryl iodides in visible-light-mediated free radical reactions

Specifications

CAS 94928-86-6
Molecular Formula C33H24IrN3
Linear Formula C33H24IrN3
MDL Number MFCD12022527
Quantity 250 mg
Synonym Ir(ppy)3; Iridium, tris[2-(2-pyridinyl-kappaN)phenyl-kappaC]
Molecular Weight (g/mol) 654.78
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.