Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris[2-phenylpyridinato-C2,N]iridium(III)

Catalog No. 694924250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-492-4250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-492-4250MG Supplier Sigma Aldrich Supplier No. 694924250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

TGA/DSC Lot specific traces available upon request

Specifications

CAS 94928-86-6
Molecular Formula C33H24IrN3
Linear Formula C33H24IrN3
Quantity 250 mg
MDL Number MFCD12022527
Synonym Ir(ppy)3; Iridium, tris[2-(2-pyridinyl-kappaN)phenyl-kappaC]
Molecular Weight (g/mol) 654.78
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.