Learn More
Medchemexpress LLC Tris(3-hydroxypropyltriazolylmethyl)amine | 760952-88-3 | MFCD27665386 | 99.0% | 434.50 g/mol | C18H30N10O3 | 250 MG
SDP
Catalog No. 5000302377
Encompass_Preferred
Tris(3-hydroxypropyltriazolylmethyl)amine is a water-soluble triazole ligand that accelerates and stabilizes copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry reactions, especially in aqueous or mixed solvent systems. Supplied as a high-purity solid, it is commonly used in bioconjugation, biorthogonal chemistry, and laboratory-scale click reaction optimization.
  • Water-soluble triazole ligand suitable for aqueous reactions.
  • Enhances copper(I)-catalyzed azide-alkyne cycloaddition reaction rates and selectivity.
  • Compatible with bioconjugation and biorthogonal chemistry workflows.
  • High purity suitable for analytical and preparative applications.
  • Stable as a powder under recommended storage conditions.
  • Available in multiple small-package sizes for laboratory use.
Catalog No. 50-003-02377 Supplier Medchemexpress LLC Supplier No. 250MGHYW021042
Tris(3-hydroxypropyltriazolylmethyl)amine is a water-soluble triazole ligand that accelerates and stabilizes copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry reactions, especially in aqueous or mixed solvent systems. Supplied as a high-purity solid, it is commonly used in bioconjugation, biorthogonal chemistry, and laboratory-scale click reaction optimization.
  • Water-soluble triazole ligand suitable for aqueous reactions.
  • Enhances copper(I)-catalyzed azide-alkyne cycloaddition reaction rates and selectivity.
  • Compatible with bioconjugation and biorthogonal chemistry workflows.
  • High purity suitable for analytical and preparative applications.
  • Stable as a powder under recommended storage conditions.
  • Available in multiple small-package sizes for laboratory use.