Learn More
Strem, An Ascensus Company CAS# 63995-70-0. 250mg. Tris(3-sulfonatophenyl)phosphine hydrate, sodium salt (<5% oxide). MFCD11046357
SDP
Supplier:  Strem, An Ascensus Company 158013250MG
Encompass
CAS# 63995-70-0. 250mg. Tris(3-sulfonatophenyl)phosphine hydrate, sodium salt (<5% oxide). MFCD11046357. Molecular Weight: 568.40. Molecular Formula: Na3P(C6H4SO3)3·;XH2O. Color/form: white to off-white pwdr. Strem# 15-8013. http://www.strem.com/catalog/v/15-8013/
Catalog No. 50-138-5439