Learn More
Cayman Chemical Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium II dichloride complex, Purity: ≥95%, CAS: 36309-88-3, 10 mg
SDP
Catalog No. 502971449
Encompass_Preferred
A luminescent probe (absorption λmax: 455 nm, luminescence λmax: 613 nm) widely used for detection and quantitation of oxygen; strongly reduced by molecular oxygen as a result of dynamic quenching; thus oxygen detection can be based on either measurement of intensity or decay time
Catalog No. 50-297-1449 Supplier Cayman Chemical Supplier No. 1611710 MG
A luminescent probe (absorption λmax: 455 nm, luminescence λmax: 613 nm) widely used for detection and quantitation of oxygen; strongly reduced by molecular oxygen as a result of dynamic quenching; thus oxygen detection can be based on either measurement of intensity or decay time