Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-biphenylyl)amine 98.0+%, TCI America™
SDP

Catalog No. T33271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33271G 1 g
T33275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33271G Supplier TCI America Supplier No. T33271G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6543-20-0
Molecular Formula C36H27N
Molecular Weight (g/mol) 473.62
MDL Number MFCD30470498
InChI Key RORPOZIIMCFMFH-UHFFFAOYSA-N
PubChem CID 18435272
IUPAC Name N,N-bis({[1,1'-biphenyl]-4-yl})-[1,1'-biphenyl]-4-amine
SMILES C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 262°C
Color White-Yellow
Quantity 1 g
Formula Weight 473.62
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-biphenylyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.