Learn More
Medchemexpress LLC Tris[4-(dimethylamino)phenyl]methanol | 467-63-0 | MFCD00071920 | 99.1% | 389.53 g/mol | C25H31N3O | 250MG
SDP
Catalog No. 5000236578
Encompass_Preferred
Solvent violet 9 (tris[4-(dimethylamino)phenyl]methanol; CAS 467-63-0) is a triarylmethane dye used for recognizing and staining DNA in laboratory applications. It is a solid with molecular formula C25H31N3O and a molecular weight of 389.5 g/mol. Typical purity is ~99.1%, and the compound is supplied in small laboratory package sizes for research use.
  • Used for recognizing and staining DNA.
  • Triarylmethane dye with strong violet color.
  • High reported purity (~99.1%).
  • Solid form, molecular weight 389.5 g/mol.
  • Supplied in multiple laboratory package sizes.
Catalog No. 50-002-36578 Supplier Medchemexpress LLC Supplier No. HY125786250MG
Add to Cart
Add to Cart
Solvent violet 9 (tris[4-(dimethylamino)phenyl]methanol; CAS 467-63-0) is a triarylmethane dye used for recognizing and staining DNA in laboratory applications. It is a solid with molecular formula C25H31N3O and a molecular weight of 389.5 g/mol. Typical purity is ~99.1%, and the compound is supplied in small laboratory package sizes for research use.
  • Used for recognizing and staining DNA.
  • Triarylmethane dye with strong violet color.
  • High reported purity (~99.1%).
  • Solid form, molecular weight 389.5 g/mol.
  • Supplied in multiple laboratory package sizes.