Learn More
Medchemexpress LLC Tris(4-methoxyphenyl)phosphine | 855-38-9 | 98.0% | 352.36 | 500 G
SDP
Catalog No. 5000454489
Encompass_Preferred
Tris(4-methoxyphenyl)phosphine is a biochemical reagent. It can be utilized as a biological material or an organic compound for life science-related research. It is intended for research use only and is not sold to patients.
  • Employed in Suzuki reaction.
  • Used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
Catalog No. 50-004-54489 Supplier Medchemexpress LLC Supplier No. HYW013772500G
Tris(4-methoxyphenyl)phosphine is a biochemical reagent. It can be utilized as a biological material or an organic compound for life science-related research. It is intended for research use only and is not sold to patients.
  • Employed in Suzuki reaction.
  • Used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.