Learn More
Medchemexpress LLC Tris(4-methoxyphenyl)phosphine | 855-38-9 | 99.3% | 352.36 | 100 G
SDP
Catalog No. 5000454636
Encompass_Preferred
Tris(4-methoxyphenyl)phosphine is a biochemical reagent used as a biological material or organic compound for life science related research. It is employed in Suzuki reaction and used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
  • Can be used as a biological material or organic compound.
  • Suitable for life science related research.
  • Employed in Suzuki reactions.
  • Used in kinetic studies of catalyst effects on biphenyl epoxy resin curing.
Catalog No. 50-004-54636 Supplier Medchemexpress LLC Supplier No. HYW013772100G
Tris(4-methoxyphenyl)phosphine is a biochemical reagent used as a biological material or organic compound for life science related research. It is employed in Suzuki reaction and used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
  • Can be used as a biological material or organic compound.
  • Suitable for life science related research.
  • Employed in Suzuki reactions.
  • Used in kinetic studies of catalyst effects on biphenyl epoxy resin curing.